missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AHNAK Polyclonal Antibody
Descripción
Immunogen sequence: MKGTKVKTPE MIIQKPKISM QDVDLSLGSP KLKGDIKVSA PGVQGDVKGP QVALKGSRVD IETPNLEGTL TGPRLGSPSG KTGTCRISMS EVDLNVAAPK VKGGVDVTLP RVEGKVKVPE VDVRGPKVDV SA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
AHNAK1 (Desmoyokin) is a large (700 kDa) scaffold protein that translocates to the plasma membrane after an increas of extracellular calcium level or upon proteinkinase C activation and regulates extracellular calcium influx mediated by L-type Ca2+ channels. AHNAK1 has been implicated in diverse signal transduction proceses affecting cell differentiation and proliferation. In response to calcium-dependent intercellular contacts AHNAK1 forms multimeric complexes in the plasma membrane, connected with actin and annexin 2/S100A10 assemblies and is thus involved in organization of the plasma membrane architecture. In epithelial cells, AHNAK1 is localized in cytoplasm or is membrane-associated, but in cells of nonepithelial origin AHNAK1 is predominantly nuclear; it has a weak DNA-binding activity and associates with the DNA-ligase IV-XRCC4 complex.
Especificaciones
Especificaciones
| Antígeno | AHNAK |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | AHNAK |
| N.º de referencia del gen | Q09666 |
| Alias de gen | AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118 |
| Símbolos de los genes | AHNAK |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?