missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGTR1 Recombinant Rabbit Monoclonal Antibody (8W6G0)

Descripción
Positive test controls include: A-549. Immunogen sequence: MILNSSTEDG IKRIQDDCPK AGRHNYIFVM IPTLYSIIFV VGIFGNSLVV IVIYFYMKLK TVASVFLLNL ALADLCFLLT LPLWAVYTAM EYRWPFGNYL.
AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.
Especificaciones
Especificaciones
| Antígeno | AGTR1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 8W6G0 |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | AGTR1 |
| N.º de referencia del gen | P30556 |
| Alias de gen | 1810074K20Rik; AG2S; AGTR1; Agtr1a; Agtr-1a; AGTR1B; AI551199; angiotensin II receptor type 1; angiotensin II receptor, type 1; angiotensin II receptor, type 1a; Angiotensin II type-1 receptor; Angiotensin II type-1A receptor; angiotensin receptor 1; angiotensin receptor 1a; Angtr-1a; AT1; At1a; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R; Type-1 angiotensin II receptor; Type-1A angiotensin II receptor; type-1B angiotensin II receptor; vascular type-1 angiotensin II receptor |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?