missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGTR1 Recombinant Rabbit Monoclonal Antibody (23GB635), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA553169
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.
Especificaciones
| AGTR1 | |
| Recombinant Monoclonal | |
| 2.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30556 | |
| AGTR1, Agtr1a | |
| A synthetic peptide of human AGTR1 (1-48AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB635 | |
| Unconjugated | |
| AGTR1 | |
| 1810074K20Rik; AG2S; AGTR1; Agtr1a; Agtr-1a; AGTR1B; AI551199; angiotensin II receptor type 1; angiotensin II receptor, type 1; angiotensin II receptor, type 1a; Angiotensin II type-1 receptor; Angiotensin II type-1A receptor; angiotensin receptor 1; angiotensin receptor 1a; Angtr-1a; AT1; At1a; AT1AR; AT1B; AT1BR; AT1R; AT2R1; AT2R1A; AT2R1B; HAT1R; Type-1 angiotensin II receptor; Type-1A angiotensin II receptor; type-1B angiotensin II receptor; vascular type-1 angiotensin II receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11607, 185 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido