missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aggrecan Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13880-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This polyclonal antibody raised against 1-298 aa of human aggrecan reveals a band of 70 kDa, which is consistent with the molecular weight of degradation product generated by proteolysis at the aggrecanase site.
Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluro binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.Especificaciones
| Aggrecan | |
| Polyclonal | |
| Unconjugated | |
| ACAN | |
| ACAN, AGC1, AGCAN, aggrecan, CSPG1, CSPGCP, MSK16, SEDK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11595, 176, 58968 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07897, P16112, Q61282 | |
| ACAN | |
| Aggrecan Fusion Protein Ag4985 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido