Learn More
ADK Mouse anti-Human, Unconjugated, Clone: 1G1A5, Proteintech
Código de producto. 16829235 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16829235 20 μL 20 microlitros
16819235 150 μL 150 microlitros
2 options
Código de producto. 16829235
Marca: Proteintech 669291IG20UL
Mouse Monoclonal Antibody
The ADK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADK. This antibody recognizes human, mouse, rat antigen. The ADK antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno ADK
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Monoclonal
Clon 1G1A5
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 132
Especies diana Human, Rat, Mouse
Isotype IgG1