missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody (MM0077-6D31), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-12014AF488
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| ADAM10 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ADAM10 | |
| Human recombinant ADAM10 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| In Western blots, this antibody shows weak cross-reactivity with recombinant mouse ADAM10 and human ADAM8, 9, 15, and 17. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0077-6D31 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Signal Transduction | |
| 102 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido