missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acylated Ghrelin Monoclonal Antibody (891240)
GREENER_CHOICE

Código de producto. 30536251
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30536251 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30536251 Proveedor Invitrogen™ N.º de proveedor MA561918

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Acylated ghrelin is a hormone primarily produced by the stomach and released into the bloodstream. It plays a significant role in regulating appetite and energy balance. Functionally, acylated ghrelin acts as an orexigenic hormone, meaning it stimulates appetite and promotes food intake. It interacts with specific receptors in the brain, particularly the growth hormone secretagogue receptor 1a (GHSR1a), to increase hunger signals and enhance food-seeking behaviors. Acylated ghrelin also influences energy homeostasis by increasing fat storage and reducing energy expenditure. Structurally, acylated ghrelin is a peptide hormone consisting of 28 amino acids. It undergoes a post-translational modification process called acylation, where a fatty acid (usually octanoic acid) is attached to the third amino acid residue (serine) in the peptide chain. This acylation is crucial for the hormone's biological activity and stability. Localization-wise, acylated ghrelin is primarily produced by the X/A-like cells in the gastric mucosa. After secretion into the bloodstream, it acts on various target tissues, including the hypothalamus, pituitary gland, and adipose tissue. The hypothalamus is particularly important for regulating appetite and energy balance, as it receives and integrates signals from acylated ghrelin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Acylated Ghrelin
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 891240
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with trehalose and no preservative
Alias de gen A-GHRL
Especie del huésped Mouse
Inmunógeno Human Ghrelin acylated peptide, Gly24-Arg51.
Método de purificación Protein A/G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.