missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACVR1 Mouse anti-Human, Clone:2C1E8, Abnova™
Mouse monoclonal antibody raised against recombinant human ACVR1.
Marca: Abnova MAB17491.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. This gene encodes activin A type I receptor which signals a particular transcriptional response in concert with activin type II receptors. Mutations in this gene are associated with fibrodysplasia ossificans progressive. [provided by RefSeq]Especificaciones
| ACVR1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| ACVR1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 2C1E8 | |
| activin A receptor, type I | |
| In PBS (0.05% sodium azide) | |
| ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI | |
| Recombinant protein corresponding to amino acid 21-120 of human ACVR1 from E. coli. | |
| 90 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido