missing translation for 'onlineSavingsMsg'
Learn More

ACTL7B (Human) Matched Antibody Pair, Abnova™

Product Code. 16193184
Change view
Click to view available options
Cantidad:
1 Set
Unit Size:
1 set
Product Code. Cantidad unitSize
16193184 1 Set 1 set
1 options
This item is not returnable. View return policy

Product Code. 16193184

Brand: Abnova H00010880AP11.1Set

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Matched Antibody Pair

Specifications

Antígeno ACTL7B
Aplicaciones ELISA
Conjugado Unconjugated
génica actin-like 7B
Alias de gen -
Símbolos de los genes ACTL7B
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10880
Incluye Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ACTL7B (100 ug) 2. Detection antibody: mouse monoclonal anti-ACTL7B, IgG2a Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Tipo de producto Ab Pair
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.