missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin (Muscle Specific) Antibody (MSA/953), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-47662C
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF
Especificaciones
| Actin (Muscle Specific) | |
| Monoclonal | |
| DyLight 650 | |
| P62736, P68133, P68032 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| MSA/953 | |
| 50 mM Sodium Borate | |
| ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
| Synthetic peptide common among human muscle actins | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers | |
| 58 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido