Descripción
ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.
Especificaciones
Especificaciones
| Antígeno | ACTH |
| Aplicaciones | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | 3C12C7 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | POMC |
| N.º de referencia del gen | P01189, P01193, P01194 |
| Alias de gen | ACTH, ACTH Melanotropin alpha, Adrenocorticotropic hormone, Beta LPH Lipotropin gamma, CLIP, CLIP Lipotropin beta, Corticotropin lipotropin, Gamma LPH Melanotropin beta, LPH, MSH, POC, POMC, Pro opiomelanocortin, proopiomelanocortin |
| Show More |