missing translation for 'onlineSavingsMsg'
Learn More

ACSL5 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene

Produktkod. 17546234
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Cantidad:
100 μL
30 μL
Förpackningsstorlek:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Cantidad unitSize
17546234 100 μL 100 microlitros
17566234 30 μL 30 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17546234 Leverantör Origene Technologies Leverantörsnummer TA811884

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in uterus and spleen, and in trace amounts in normal brain, but has markedly increased levels in maligt gliomas. This gene functions in mediating fatty acid-induced glioma cell growth. Three transcript variants encoding two different isoforms have been found for this gene.

Specifikationer

Antígeno ACSL5
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI1C6
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica ACSL5
N.º de referencia del gen Q8JZR0, Q9ULC5
Alias de gen ACS2, ACS5, FACL5
Símbolos de los genes Acsl5
Especie del huésped Mouse
Inmunógeno Human recombit protein fragment corresponding to amino acids 104-620 of human ACSL5 produced in E.coli.
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 433256, 51703
Especies diana Human, Mouse
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.