Learn More
ACP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11594-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher′s disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin′s lymphoma and in the sera of patients undergoing active bone turnover.Especificaciones
| ACP5 | |
| Polyclonal | |
| Unconjugated | |
| ACP5 | |
| ACP5, Tartrate resistant acid ATPase, TR AP, TRACP, TRAP, TrATPase, Type 5 acid phosphatase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11433, 25732, 54 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13686, P29288, Q05117 | |
| ACP5 | |
| ACP5 Fusion Protein Ag2065 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |