missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: YRGGYFSGEQ AGEVLESAVL ALCSQLKDDA VALVDVIAPP DFVLDSPIGR ADGELYKNLW GAVLQESKVL ERASWWPEFS VNKPVIGSLK SKL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
Acyl-Coenzyme A oxidase 3 also know as pristanoyl-CoA oxidase (ACOX3) is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined.
Especificaciones
Especificaciones
| Antígeno | ACOX3 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | ACOX3 |
| N.º de referencia del gen | O15254 |
| Alias de gen | Acox3; acyl-CoA oxidase 3, pristanoyl; acyl-Coenzyme A oxidase 3, pristanoyl; BI685180; branched-chain acyl-CoA oxidase; BRCACox; BRCOX; EST-s59; PCOX; Peroxisomal acyl-coenzyme A oxidase 3; PRCOX; Pristanoyl-CoA oxidase |
| Símbolos de los genes | ACOX3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
