missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Acetyl-HIST1H2BC (Lys108) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5144328
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Acetyl-HIST1H2BC (Lys108) Polyclonal Antibody for ICC/IF, ELISA, ChIP
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. The protein has antibacterial and antifungal antimicrobial activity. The main transcript variant of this gene is intronless and encodes a replication-dependent histone that is a member of the histone H2B family. This transcript variant lacks a polyA tail but instead contains a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Especificaciones
| Acetyl-HIST1H2BC (Lys108) | |
| Polyclonal | |
| Unconjugated | |
| H2BC5 | |
| dJ221C16.3; H2B Clustered Histone 4; H2B histone family, member L; H2B.1; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bc; histone cluster 1, H2bc; histone H2B type 1-C/E/F/G/I; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3017 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, ELISA, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P58876, P62807 | |
| H2BC5, Hist1h2bc | |
| Peptide sequence around site of Acetyl-Lys (108) derived from Human Histone H2B type 1-C/E/F/G/I. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido