missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Pure Grade, Penta Chemicals

Código de producto. 30020601
Change view
Click to view available options
Cantidad:
1 L
2.5 L
Tamaño de la unidad:
1 litro
2.50 litro
Código de producto. Cantidad unitSize
30020601 2.5 L 2.50 litro
30020484 1 L 1 litro
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30020601

Marca: PENTA CHEMICALS 1997012500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Identifiants chimiques

CAS 64-19-7
Peso molecular (g/mol) 60.05

Spécification

Nombre del producto químico o material Acetic Acid
Porcentaje de pureza ≥99%
Grado Pure
Cantidad 2.5 L
Envase Glass Bottle
compliance-icons compliance-icons
Identifiant produit
  • Acetic acid 99%
Mention d'avertissement
  • Peligro
Catégorie de danger
  • Lesión ocular grave/irritación ocular Categoría 1
  • Líquido inflamable Categoría 3
  • Corrosión o irritación cutáneas Categoría 1A
Mention de danger
  • H226-Líquidos y vapores inflamables.
  • H314-Provoca quemaduras graves en la piel y lesiones oculares graves.
  • H318-Provoca lesiones oculares graves.
Conseils de prudence
  • P260-No respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
  • P280-Llevar guantes/prendas/gafas/máscara de protección.
  • P301+P330+P331-EN CASO DE INGESTIÓN: Enjuagarse la boca. NO provocar el vómito.
  • P305+P351+P338-EN CASO DE CONTACTO CON LOS OJOS: Aclarar cuidadosamente con agua durante varios minutos. Quitar las lentes de contacto, si lleva y resulta fácil. Seguir aclarando.
  • P310-Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.
Informations supplémentaires
  • MIXTURE LIST-Contém: Acetic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.