missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACBD7 Monoclonal Antibody (08), Invitrogen™
Description
Specificity: Human ACBD7
Binds medium- and long-chain acyl-CoA esters.
Specifications
Specifications
| Antígeno | ACBD7 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 08 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | ACBD7 |
| N.º de referencia del gen | Q8N6N7 |
| Alias de gen | 9230116B18Rik; ACBD7; acyl-CoA binding domain containing 7; acyl-CoA-binding domain-containing protein 7; acyl-Coenzyme A binding domain containing 7; bA455B2.2; RGD1564164 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?