missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABHD4 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15729380 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15729380 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15729380

Marca: Invitrogen™ PA551421

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: ADDLEQQSQG WLSSWLPTWR PTSMSQLKNV EARILQCLQN KFLARYVSLP NQNKIWTVTV SPEQNDRTPL VMVHGFGGGV GLWILNMDSL SARRTLHTFD LLGFGRSSRP AFPRDPEGAE DEFVTSIETW RETMGIPSMI LLGHSLGGF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The α/β hydrolase superfamily is comprised of diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical α/β hydrolase fold, which consists of an eight-stranded parallel α/β structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. ABHD1 plays a role in metabolizing smoking xenobiotics. ABHD2 participates in the development of atherosclerosis. ABHD4 is involved in an alternative synthesis pathway of NAE. ABHD4 is a lysophospholipase selective for N-acyl phosphatidylethanolamine (NAPE) which participates in the biosynthesis of N-acyl ethanolamines. Mutations in ABHD5 contribute to Chanarin-Dorfman syndrome. ABDH6 may play a role in nervous system metabolism and signaling. ABHD14A is possibly involved in granule neuron development.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ABHD4
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica ABHD4
N.º de referencia del gen Q8TB40
Alias de gen (Lyso)-N-acylphosphatidylethanolamine lipase; (Lyso)-N-acylphosphatidylethanolamine lipase; protein ABHD4; 1110035H23Rik; Abh4; Abhd4; abhydrolase domain containing 4; abhydrolase domain containing 4, N-acyl phospholipase B; abhydrolase domain-containing protein 4; AI429574; Alpha/beta hydrolase domain-containing protein 4; alpha/beta-hydrolase 4; hypothetical protein LOC509896; lyso-N-acylphosphatidylethanolamine lipase; protein ABHD4
Símbolos de los genes ABHD4
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human ABHD4. Recombinant protein control fragment (Product #RP-88857).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 63874
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.