missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Abciximab Chimeric Recombinant Human Monoclonal Antibody

Descripción
Purity >95% Endotoxin level <10EU/mg by LAL method. Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Abciximab is a Fab fragment of the chimeric human-murine monoclonal antibody 7E3. Abciximab binds to the glycoprotein (GP) IIb/IIIa receptor of human platelets and inhibits platelet aggregation by preventing the binding of fibrinogen, von Willebrand factor, and other adhesive molecules. It also binds to vitronectin (alpha-v-beta-3) receptor found on platelets and vessel wall endothelial and smooth muscle cells. Abciximab binds to the intact platelet GpiIb/IIIa receptor, which is a member of the integrin family of adhesion receptors and the major platelet surface receptor involved in platelet aggregation. This binding is thought to involve steric hindrance and/or conformational alterations which block access of large molecules to the receptor rather than direct interaction with the RGD (arginine-glycine-aspartic acid) binding site of GpiIb/IIIa. By binding to the vitronectin receptor (also known as the alpha-v-beta-3 integrin), abciximab blocks effects mediated by this integrin which include cell adhesion. Furthermore, abciximab blocks Mac-1 receptor on monocytes and neutrophils thus inhibiting monocyte adhesion.
Especificaciones
Especificaciones
| Antígeno | Abciximab Chimeric |
| Aplicaciones | ELISA |
| Clasificación | Recombinant Monoclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.5 |
| Alias de gen | c7E3 |
| Especie del huésped | Human |
| Inmunógeno | Human ITGA2B_ITGB3. |
| Método de purificación | Protein L, Protein A/G |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?