Learn More
AADACL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16852484 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16852484 20 μL 20 microlitros
16842484 150 μL 150 microlitros
2 options
Código de producto. 16852484
Marca: Proteintech 140211AP20UL
Rabbit Polyclonal Antibody
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.
Especificaciones
Antígeno AADACL1
Aplicaciones Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.18 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NCEH1
N.º de referencia del gen B2GV54, Q6PIU2, Q8BLF1
Alias de gen AADACL1, KIAA1363, KIAA1363, NCEH1,NCEH, NCEH, NCEH1
Símbolos de los genes Nceh1
Especie del huésped Rabbit
Inmunógeno AADACL1 Fusion Protein Ag5136
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 294930, 320024, 57552
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG