Learn More
A2BP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16822025 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16822025 150 μL 150 microlitros
16832025 20 μL 20 microlitros
2 options
Código de producto. 16822025
Marca: Proteintech 226471AP150UL
Rabbit Polyclonal Antibody
This is a rabbit polyclonal antibody raised against the N terminus of human A2BP1.
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.
Especificaciones
Antígeno A2BP1
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.27 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica RBFOX1
N.º de referencia del gen Q9JJ43, Q9NWB1
Alias de gen A2BP, A2BP1, ataxin 2 binding protein 1, Fox 1 homolog A, FOX1, HRNBP1, RBFOX1
Símbolos de los genes RBFOX1
Especie del huésped Rabbit
Inmunógeno A2BP1 Fusion Protein Ag18527
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 268859, 54715
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG