Learn More
Tuttnauer A114 Gap Filler
Tienda Tuttnauer Productos
Click to view available options
:
Gap Filler; For use with: Tiva Washer
Descripción
- A114 Hexagon head screw with M8x1 thread To be screwed to the positions of the rack where the Nozzles are not placed, to allow the passage of the washing water only in the occupied positions.
Especificaciones
Especificaciones
| Material | Acero inoxidable |
| Tipo de producto | Nozzle |
| Para utilizar con (equipo) | Tiva Washer |