Learn More
Tygon A-60-G Chemical Dispensing Tubing
Código de producto. p-200105035 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/8 in
1 in
3/16 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
Diámetro exterior (imperial):
1-1/4 in
1/2 in
1/4 in
11/16 in
13/16 in
1 in
3/16 in
3/8 in
5/16 in
5/8 in
7/16 in
7/8 in
9/16 in
Tamaño de la unidad:
15 metros
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30054342 5/16 in 7/16 in 15 metros
30054000 5/8 in 13/16 in 15 metros
30053254 5/8 in 7/8 in 15 metros
30053593 1/8 in 1/4 in 15 metros
30053538 1/2 in 11/16 in 15 metros
30054173 3/8 in 1/2 in 15 metros
30053776 3/8 in 5/8 in 15 metros
30053394 1/16 in 3/16 in 15 metros
30053266 5/16 in 1/2 in 15 metros
30053430 1/4 in 3/8 in 15 metros
30053152 3/16 in 5/16 in 15 metros
30053567 1/2 in 5/8 in 15 metros
30054367 1 in 1-1/4 in 15 metros
30053761 1/4 in 1/2 in 15 metros
30054059 7/16 in 9/16 in 15 metros
30053314 3/4 in 1 in 15 metros
30053479 3/8 in 9/16 in 15 metros
30054091 1/8 in 3/8 in 15 metros
30053424 1/4 in 7/16 in 15 metros
19 options
Código de producto. 30054342
Marca: Tygon 5010657
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Specially formulated tubing offers excellent resistance to inorganic fluids
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubing
Especificaciones
Diámetro interior (imperial) 5/16 in
Material Tygon A-60-G
Diámetro exterior (imperial) 7/16 in
Longitud global (métrico) 50 ft.
Color Negro