Learn More
DiscoverX 9-cis-Retinoic acid
Código de producto. 17302469
Encompass
Change view
Click to view available options
:
1 mg
Código de producto.
17302469 1 mg
1 options
Código de producto. 17302469
Marca: DiscoverX 921136
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
9-cis-Retinoic acid is an agonist for retinoic acid and retinoid X receptors RARα, RARβ, RXRα, and RXRγ. Provided as 1mg of dry powder.
9-cis-Retinoic acid is a natural metabolite of vitamin A (retinol) and potent agonist of the retinoic acid and retinoid X receptors (Ki=0.5-27nM and 3.8-12nM, respectively). Mediates retinol functions for growth and development, and is active in embryonic development by intercellular signaling for the posterior portion of the embryo, and developmental patterning. Provided as 1mg of dry powder.
Especificaciones
Cantidad 1 mg