missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Regulator that plays a central role in regulation of apoptosis and cell growth via its interactions. Regulates TP53 by enhancing the DNA binding and transactivation function of TP53 on the promoters of proapoptotic genes in vivo. Inhibits the ability of APPBP1 to conjugate NEDD8 to CUL1, and thereby decreases APPBP1 ability to induce apoptosis. Impedes cell cycle progression at G2/M. Its apoptosis-stimulating activity is inhibited by its interaction with DDX42.
Specifications
Specifications
| Antígeno | 53BP2 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | TP53BP2 |
| N.º de referencia del gen | Q13625 |
| Alias de gen | 53BP2, ASPP2, BBP, p53 binding protein 2, p53BP2, PPP1R13A, TP53BP2 |
| Símbolos de los genes | TP53BP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?