Learn More
Atropine methyl bromide, MedChemExpress
Código de producto. 16490857
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16490857 50 mg 50 miligramos
16485406 100 mg 100 miligramos
16464558 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16490857 Proveedor Medchem Express N.º de proveedor HY112076
MedChemExpress Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine.
Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine.
Especificaciones
Nombre del producto químico o material Atropine methyl bromide
Forma física Solid
CAS 2870-71-5
Color Off-White
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C18H26BrNO3
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Sinónimo Methylatropine bromide
Información de solubilidad DMSO : 150 mg/mL (390.31 mM; Need ultrasonic)
SMILES C[N+]1([C@H]2C[C@H](OC(C(C3=CC=CC=C3)CO)=O)C[C@@H]1CC2)C.[Br-]
Peso molecular (g/mol) 384.31
Formula Weight (peso de la fórmula) 384.31
Porcentaje de pureza 95.0%