Learn More
AICAR phosphate, MedChemExpress
Descripción
AICAR phosphate (Acadesine phosphate) is an adenosine analog and a AMPK activator. AICAR phosphate regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR phosphate is also an autophagy, YAP and mitophagy inhibitor.
Especificaciones
Especificaciones
| Nombre del producto químico o material | AICAR phosphate |
| CAS | 681006-28-0 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C9H17N4O9P |
| Cantidad | 50 mg |
| Sinónimo | Acadesine phosphate AICA Riboside phosphate |
| Información de solubilidad | H2O : 100 mg/mL (280.72 mM; Need ultrasonic) |
| SMILES | OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O1.O=P(O)(O)O |
| Peso molecular (g/mol) | 356.23 |
| Mostrar más |