missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Standard: Cr @ 5000 μg/g in Hydrocarbon Oil, VHG™
Click to view available options
Cantidad:
50 g
200 g
Tamaño de la unidad:
200 g
50 g
Description
Chromium Standard: Cr @ 5000 μg/g in Hydrocarbon Oil
Specifications
Specifications
| Nombre del producto químico o material | Chromium |
| CAS | 7440-47-3 |
| Métodos | ICP/XRF/RDE |
| Nombres de analitos o componentes | Chromium |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | Cr |
| Concentración | 5000 μg/g |
| Certificaciones/conformidad | ISO 17034 |
| Sinónimo | Cr |
| Fórmula InChI | InChI=1S/Cr |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction