missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 500 kDa FITC-dextran Sulfate Green Fluorophore
Descripción
- Product Overview: FITC-dextran sulfates are highly sulfated dextran molecules covalently labeled with fluorescein isothiocyanate (FITC), combining strong negative charge with bright green fluorescence. Available in 4 kDa, 10 kDa, 40 kDa, and 500 kDa molecular weights, FITC-dextran Sulfates are ideal for permeability studies, barrier function assays, and molecular interaction research where anionic tracers are required.
- Features:
- Green Fluorescence: Excitation maximum at 495 nm and emission maximum at 519 nm, compatible with standard FITC filter sets.
- Highly Sulfated Structure: Provides strong negative charge density, mimicking the behavior of naturally occurring polyanions like heparin.
- Wide Molecular Weight Range: Offered at 4 kDa, 10 kDa, 40 kDa, and 500 kDa to match different permeability and diffusion model requirements.
- Water-Soluble: Fully soluble in aqueous media, facilitating easy preparation for biological and analytical experiments.
- Controlled Labeling and Consistency: Manufactured under strict specifications to ensure reproducible fluorescence intensity and sulfation degree across batches.
- Benefits:
- Reliable Permeability Probes: Ideal for studying transport properties across biological membranes, vascular barriers, and engineered tissues.
- Electrostatic Interaction Research: Useful for investigating binding to cationic proteins, peptides, or biomaterials.
- Modeling Charged Molecule Behavior: Acts as an analog for anionic macromolecules in studies of transport, adsorption, and separation.
- Flexible Application Across Systems: Suitable for live-cell, tissue culture, microfluidics, and in vivo tracer studies.
- Bright and Stable Fluorescence: Enables sensitive detection and quantitative analysis using fluorescence-based methods.
- Applications:
- Barrier Function Assays: Used to assess the integrity of epithelial and endothelial monolayers, including gut and blood-brain barrier models.
- Vascular Permeability Studies: Applied to measure vascular leakiness under physiological and pathological conditions.
- Charge-Dependent Binding Studies: Investigates interactions between anionic molecules and cationic surfaces or proteins.
- Tracer Studies in Microfluidics and Perfusion Systems: Tracks the movement of charged molecules through engineered or natural channels.
- Cell Culture and Tissue Engineering Models: Maps molecular diffusion and barrier formation in tissue constructs and organoids.
- Technical Specifications:
- Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm.
- Molecular Weights: 4 kDa, 10 kDa, 40 kDa, and 500 kDa.
- Appearance: Light-sensitive, greenish dry powder, readily soluble in water.
- Sulfation Level: High, typically 16 to 20% sulfur content, depending on grade.
- Storage and Stability:
- Store dry, protected from light, at 2°C to 8°C for maximum stability and fluorescence preservation.
- Stable under recommended conditions for long-term storage and experimental reliability.
- Packaging and Availability:
- Supplied as dry powder, facilitating long shelf life and ease of storage.
- Available in various pack sizes to support both small-scale experiments and larger research projects.
Especificaciones
Especificaciones
| Cantidad | 100 mg |
| Tipo de producto | Fluorescent Labeling Reagent |
| Peso molecular | 500 kDa |
| Tipo de etiqueta | Other Labels or Dyes |
| Color | Green |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?