Learn More
5-Cloro-1-pentanol, 95 %, Thermo Scientific Chemicals
Description
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
El 5-cloro-1-pentanol se utiliza para producir 6-hidroxi-hexanitrilo a una temperatura de 90 a 100 °C. También es una importante materia prima y se utiliza como intermedio en la síntesis orgánica, los productos farmacéuticos y los agroquímicos.
Solubilidad
Ligeramente soluble en agua.
Notas
Mantener el recipiente bien cerrado. Almacenar en un lugar fresco y seco y en recipientes bien sellados.
Specifications
Specifications
| Nombre del producto químico o material | 5-Chloro-1-pentanol |
| Densidad | 1.06 |
| Punto de ebullición | 102°C to 103°C (8 mmHg) |
| Índice de refracción | 1.454 |
| Cantidad | 10 g |
| Beilstein | 1732314 |
| Información de solubilidad | Slightly soluble in water. |
| Formula Weight (peso de la fórmula) | 122.6 |
| Porcentaje de pureza | 95% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.