missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminolevulinic acid hydrochloride, MedChemExpress
Description
5-Aminolevulinic acid hydrochloride (5-ALA hydrochloride) is an intermediate in heme biosynthesis in the body and the universal precursor of tetrapyrroles.
Specifications
Specifications
| Nombre del producto químico o material | 5-Aminolevulinic acid hydrochloride |
| CAS | 5451-09-2 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C5H10ClNO3 |
| Cantidad | 5 g |
| Sinónimo | 5-ALA hydrochlorideδ-Aminolevulinic acid hydrochloride5-Amino-4-oxopentanoic acid hydrochloride |
| Información de solubilidad | DMSO : 100 mg/mL (596.69 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (99.47 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(O)CCC(CN)=O.[H]Cl |
| Peso molecular (g/mol) | 167.59 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?