missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 5(6)-Aminofluorescein
Specifications
Specifications
| Cantidad | 500 mg |
| Tipo de producto | Dye Reagents |
| Peso molecular | 347.32 g/mol |
| Solubilidad | DMSO : ≥ 39 mg/mL (112.29 mM) |
| Contenido y almacenamiento | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Condiciones de envío | Room temperature |
| Objetivo | HBV |
| CAS | 27599-63-9 |
| Sinónimo | 5(6)-AFM ∣ Fluoresceinamine mixed isomers |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?