Learn More
SHOWA™ 451 Series Natural Rubber Palm Coated Thermal-Insulated Gloves
Descripción
Benifits
- Thermal Insulation
- Flexible
- Breathable
- Extra Grip
- Seamless Knit
- Skin-friendly
- Abrasion-resistant
Features
- Rough Grip
- Ergonomic
- Elasticated Wrist Cuff
- Natural Rubber Palm Coating
Trades & Applications
- Signalers
- Brick Laying
- Carpentry
- Driving Machines
- Masonry
- Demolition
- Shipping/Receiving
Especificaciones
Especificaciones
| Certificaciones/conformidad | CE Cat II, EN 388:2016, EN 511:2006 |
| Área revestida | Palm |
| Estructura | Seamless Knit |
| Coating Material | Natural Rubber |
| Color | Gris |
| Material | Cotton, Polyester |
| Tamaño | XL |
| Material de revestimiento | Acrílico |
| Resistente a | Abrasion Resistant |
| Serie | 451 |