Learn More
4-Bencil-3-tiosemicarbazida, + 98 %, Thermo Scientific Chemicals
Código de producto. p-7045489
Change view
Click to view available options
Cantidad:
1 g
5 g
Unit Size:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
11313076 1 g 1 g
11323076 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313076 Supplier Thermo Scientific Alfa Aesar Supplier No. L10920.03
Solicitar formato a granel o personalizado
This item is not returnable. View return policy
CAS: 13431-41-9 | C8H11N3S | 181.257 g/mol
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La 4-bencil-3-tiosemicarbazida se utiliza como intermedio farmacéutico.
Solubilidad
Insoluble en agua.
Notas
Almacenar en un lugar fresco. Mantener el recipiente bien cerrado en un lugar seco y bien ventilado. Mantener alejado de los agentes oxidantes fuertes. Estable en las condiciones de almacenamiento recomendadas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13431-41-9
Fórmula molecular C8H11N3S
Peso molecular (g/mol) 181.257
Número MDL MFCD00025145
Clave InChI ZTRUHAVBRPABTK-UHFFFAOYSA-N
Sinónimo 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
PubChem CID 737135
Nombre IUPAC 1-amino-3-benciltiourea
SMILES C1=CC=C(C=C1)CNC(=S)NN
Specifications
Nombre del producto químico o material 4-Benzyl-3-thiosemicarbazide
Punto de fusión 128°C to 131°C
Cantidad 1 g
Número UN UN2811
Beilstein 2096431
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 181.26
Porcentaje de pureza ≥98%