missing translation for 'onlineSavingsMsg'
Learn More

4-1BB Ligand/TNFSF9 Antibody (203942), Janelia Fluor™ 646, Novus Biologicals™

Código de producto. 18093284
Change view
Click to view available options
Cantidad:
0.5 mL
Unit Size:
0.50 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18093284 0.5 mL 0.50 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093284 Supplier Bio-Techne Supplier No. FAB1246J

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno 4-1BB Ligand/TNFSF9
Aplicaciones Western Blot, Flow Cytometry
Clasificación Monoclonal
Clon 203942
Conjugado Janelia Fluor 646
Formulación 50mM Sodium Borate
Alias de gen 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Especie del huésped Rat
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9, Arg104-Glu309, Accession # P41274
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.5 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Biologically Active Proteins
Primario o secundario Primary
ID de gen (Entrez) 8744
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.