missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 4-1BB Ligand Recombinant Rabbit Monoclonal Antibody (8E5D6)
Tienda Thermo Scientific Productos

Descripción
Positive Samples: HeLa, Raji, SH-SY5Y, Mouse lung, Mouse brain, Rat lung Immunogen sequence: GEGSGSVSLA LHLQPLRSAA GAAALALTVD LPPASSEARN SAFGFQGRLL HLSAGQRLGV HLHTEARARH AWQLTQGATV LGLFRVTPEI PAGLPSPRSE.
CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
Especificaciones
Especificaciones
| Antígeno | 4-1BB Ligand |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 8E5D6 |
| Concentración | 0.8 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | TNFSF9 |
| N.º de referencia del gen | P41273, P41274 |
| Alias de gen | 4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?