Learn More
STEMCELL Technologies 35 mm Culture Dishes
Descripción
- 35 mm culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays
- They are manufactured from optical quality, thermally resistant, non-pyrogenic, non-toxic polystyrene
- Recommended for: Hematopoietic progenitor cultures in methylcellulose-based media (MethoCult™)
- Embryoid body formation from mouse embryonic and induced pluripotent stem cells
- Performing the neural colony-forming cell assay (NCFC)
Especificaciones
Especificaciones
| Para utilizar con (aplicación) | Cell Culture; Colony Assay |
| Cantidad | 10 placas |