Learn More
DiscoverX Ready-to-Assay™ CCR3 Chemokine Receptor Frozen Cells
Click to view available options
Cantidad:
2 viales de células
Tamaño de la unidad:
2 ampollas
Descripción
The Ready-to-Assay™ CCR3 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. TARC, MDC) based activation of CCR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress CCR3 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CCR3, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Especificaciones
Especificaciones
| Entrada del ensayo | CCR3 |
| Formato | 2 Vials of Cells |
| Para utilizar con (aplicación) | Frozen Cells |
| Agonista primario | TARC, MDC |
| Cantidad | 2 viales de células |
| Entrada diana | CCR3 |