missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, Mikromol™
Description
(5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone
Specifications
Specifications
| Nombre del producto químico o material | (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone |
| CAS | 496031-57-3 |
| Nombres de analitos o componentes | (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | C14 H16 Cl F N2 O3 |
| Certificaciones/conformidad | ISO 17025 |
| Sinónimo | 9-Allylidene-2-(trifluoromethyl)thioxanthene 10-Oxide, 2-Oxazolidinone, 5-(chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-, (5R)-, (5R)-5-(Chloromethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone |
| Fórmula InChI | InChI=1S/C14H16ClFN2O3/c15-8-11-9-18(14(19)21-11)10-1-2-13(12(16)7-10)17-3-5-20-6-4-17/h1-2,7,11H,3-6,8-9H2/t11-/m0/s1 |
| SMILES | Fc1cc(ccc1N2CCOCC2)N3C[C@H](CCl)OC3=O |
| Nombre IUPAC | (5R)-5-(chloromethyl)-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?