missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethinylestradiol, Mikromol™
Descripción
Ethinylestradiol
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ethinylestradiol |
| CAS | 57-63-6 |
| Nombres de analitos o componentes | Ethinylestradiol |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | C20 H24 O2 |
| Certificaciones/conformidad | ISO 17034 |
| Sinónimo | (17alpha)-19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol, 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,17-diol, 17-Ethinyl-3,17-estradiol, 17-Ethinylestradiol, 17-Ethynyl-3,17-dihydroxy-1,3,5-oestratriene, 17-Ethynylestra-1,3,5(10)-triene-3,17beta-diol, 17-Ethynylestradiol, 17-Nor-17alpha-pregna-1,3,5-(10)-trien-20-yne-3,17-diol, 17alpha-Ethinyl-1,3,5(10)-estratriene-3,17-diol, 17alpha-Ethinyl-17beta-estradiol, 17alpha-Ethinyl-3,17-dihydroxy-delta1,3,5-estratriene, 17alpha-Ethinylestra-1,3,5(10)-triene-3,17beta-diol, 17alpha-Ethinylestradiol, 17alpha-Ethynylestra-1,3,5(10)-triene-3,17beta-diol, 17alpha-Ethynylestradiol, 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,17beta-diol, Amenoron, Chee-O-Gen, Chee-O-Genf, Diogyn E, Dyloform, Esteed, Estigyn, Estinyl, Eston-E, Estoral, Estorals, 17-Ethynylestradiol, Ethidol, Ethinoral, Ethinylestradiol, Ethinyloestradiol, Ethynylestradiol, Ethynyloestradiol, Eticyclin, Eticyclol, Etinestrol, Etinestryl, Etinoestryl, Etistradiol, Follicoral, Ginestrene, Inestra, Linoral, Lynoral, Menolyn, Microfollin, NSC 10973, Novestrol, Oradiol, Orestralyn, Palonyl, Perovex, Primogyn, Primogyn C, Primogyn M, Progynon C, Prosexol, Spanestrin, neo-Estrone |
| Fórmula InChI | InChI=1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C |
| Nombre IUPAC | (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?