missing translation for 'onlineSavingsMsg'
Learn More
Learn More
200.7 ICP Tuning Solution: Cu, Pb @ 10 μg/mL in 5% HNO3, VHG™
Click to view available options
Cantidad:
100 mL
500 mL
Tamaño de la unidad:
100 ml
500 ml
Beskrivning
200.7 ICP Tuning Solution: Cu, Pb @ 10 μg/mL in 5% HNO3
Specifikationer
Specifikationer
| Nombre del producto químico o material | Copper/Lead |
| CAS | 7440-50-8, 7439-92-1 |
| Métodos | ICP/ICP-MS |
| Nombres de analitos o componentes | Copper/Lead |
| Condiciones de envío | Room Temperature |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Cantidad | 500 mL |
| Concentration or Composition (by Analyte or Components) | Copper 10 μg/mL/Lead 10 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction