Learn More
DiscoverX 2-Methylthio-ATP
Código de producto. 17302539
Encompass
Change view
Click to view available options
Cantidad:
10 mg
Tamaño de la unidad:
10 miligramos
17302539 10 mg 10 miligramos
1 options
Código de producto. 17302539
Marca: DiscoverX 921214
2-Methylthio-ATP is an agonist for the P2γ purinergic receptor. Provided as 10mg of dry powder.
2-(Methylthio)adenosine 5′-triphosphate (2-methylthio-ATP) is an ATP analog that is a more selective agonist of the P2γ purinergic receptor. Adenosine 5'-triphosphate is a nucleoside triphosphate that is key to intracellular energy transfer. It is responsible for transporting chemical energy within cells for metabolism, is an end product of phosphorylation, cellular respiration, and fermentation, and is used in many cellular processes. Provided as 10mg of dry powder.
Especificaciones
Tipo de producto Control Ligands
Cantidad 10 mg