Learn More
2,3,5-Tri-O-bencil-D-ribofuranosa, 98 %, Thermo Scientific Chemicals
Descripción
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Se utiliza como intermedio farmacéutico. Los benzoiltiofenos son potenciadores alostéricos (AE) de la actividad agonista en el receptor A1 de la adenosina.
Solubilidad
Poco soluble en agua.(0,26 g/l) (25 °C),
Notas
Almacenar en un lugar fresco y seco. Garantizar una ventilación adecuada. Incompatible con agentes oxidantes.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2,3,5-Tri-O-benzyl-D-ribofuranose |
| Cantidad | 1 g |
| Información de solubilidad | Sparingly soluble in water.(0.26g/L) (25°C), |
| Formula Weight (peso de la fórmula) | 420.5 |
| Porcentaje de pureza | 98% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.