missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard: S @ 15 μg/g (0.0015 wt%) in #2 Diesel Fuel, VHG™

Código de producto. 17921896
Change view
Click to view available options
Cantidad:
100 mL
500 mL
1 L
1 gal
Unit Size:
1 litro
100 ml
500 ml
Product Code. Cantidad unitSize
17921896 1 L 1 litro
15707286 100 mL 100 ml
17974016 500 mL 500 ml
17982486 1 gal -
4 options
This item is not returnable. View return policy

Product Code. 17921896

Brand: VHG Labs VHGSDSL151L

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur Standard: S @ 15 μg/g (0.0015 wt%) in #2 Diesel Fuel
TRUSTED_SUSTAINABILITY

Specifications

Nombre del producto químico o material Sulfur
CAS 7704-34-9
Métodos XRF
Nombres de analitos o componentes Sulfur
Condiciones de envío Room Temperature
Fórmula molecular S
Certificaciones/conformidad ISO 17034
Fórmula InChI InChI=1S/S
SMILES [S]
Nombre IUPAC sulfur
Solvent or Matrix #2 Diesel Fuel
Peso molecular (g/mol) 32.065
Cantidad 1 L
Formula Weight (peso de la fórmula) 31.972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.