Learn More
Keystone Cleanroom Products Cubierta de equipo elástica Tyvek™ 1422A
GREENER_CHOICE
Código de producto. p-7203255 Tienda Keystone Cleanroom Products Productos
Change view
Click to view available options
Longitud (imperial):
20 in
28 in
30 in
32 in
34 in
36 in
42 in
44 in
48 in
50 in
54 in
56 in
60 in
64 in
74 in
Anchura (imperial):
16 in
20 in
22 in
24 in
26 in
28 in
30 in
32 in
34 in
36 in
40 in
44 in
50 in
54 in
78 in
Altura (imperial):
5 in
8 in
12 in
14 in
16 in
30 in
40 in
42 in
44 in
50 in
54 in
60 in
72 in
78 in
86 in
Tamaño de la unidad:
paquete de 25
paquete de 50
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Longitud (imperial) Anchura (imperial) Altura (imperial) unitSize
17778267 20 in 20 in 8 in paquete de 50
17708277 28 in 28 in 5 in paquete de 50
17718277 28 in 28 in 50 in paquete de 50
17728277 30 in 16 in 16 in paquete de 50
17738277 30 in 30 in 40 in paquete de 50
17748277 30 in 22 in 12 in paquete de 50
17708287 32 in 32 in 60 in paquete de 50
17738287 34 in 34 in 42 in paquete de 50
17748287 36 in 26 in 60 in paquete de 50
18066991 42 in 32 in 16 in paquete de 50
17778287 42 in 34 in 60 in paquete de 50
17718297 44 in 44 in 60 in paquete de 50
17738297 48 in 24 in 30 in paquete de 50
17728307 50 in 50 in 14 in paquete de 50
17768307 54 in 54 in 86 in paquete de 50
17778307 56 in 36 in 60 in paquete de 50
17788307 60 in 28 in 54 in paquete de 50
17708317 60 in 30 in 72 in paquete de 50
17718317 60 in 40 in 54 in paquete de 50
17738317 60 in 44 in 44 in paquete de 50
17758317 60 in 50 in 60 in paquete de 50
17778317 60 in 78 in 60 in paquete de 50
17788317 64 in 24 in 78 in paquete de 25
17798317 74 in 26 in 86 in paquete de 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Código de producto. 17778267 Proveedor Keystone Cleanroom Products N.º de proveedor COVTVK201608E
Solo quedan null
Añadir a la cesta
Añadir a la cesta
1422A Tyvek™ Elastic Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
  • Mantiene el equipo limpio y protegido de la contaminación
  • Transpirable
  • Ajuste de forma, rápido y uniforme
  • Fabricados y envasados en un entorno de sala limpia ISO 6
  • Embalaje práctico: 1 bolsa de polietileno sellada con calor
  • Certificados de conformidad suministrados con el envío
Especificaciones
Esterilizables en autoclave Autoclavable
Certificaciones/conformidad COC
Tipo de tapón None
Color Blanco
Desechables Reusable
Material Tyvek
Esterilidad Non-sterile
Sin látex Yes
Dimensiones (Largo x Ancho x Alto) 20 x 16 x 8 in
Longitud (imperial) 20 in
Anchura (imperial) 20 in
Altura (imperial) 8 in
Altura (métrico) 20.3 cm
Tipo de producto Cover
Longitud (métrico) 50.8 cm
Anchura (métrico) 50.8 cm