missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 14-3-3 gamma Recombinant Rabbit Monoclonal Antibody (23GB1970), Invitrogen™
GREENER_CHOICE

Código de producto. p-200087792
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30089431 100 μL 100 microlitros
30089763 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30089431 Proveedor Invitrogen™ N.º de proveedor MA553186

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

14-3-3 gamma encodes a product that belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno 14-3-3 gamma
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 23GB1970
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica YWHAG
N.º de referencia del gen P61981, P61982, P61983
Alias de gen 14-3-3 gamma; 14-3-3 protein gamma; 14-3-3 protein gamma subtype; 14-3-3 protein gamma, N-terminally processed; 14-3-3 protein gamma-1; 14-3-3 protein gamma-subtype; 14-3-3G; 14-3-3gamma; 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; 3-monooxgenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide 1; cb47; D7Bwg1348e; fd20c02; hypothetical protein; KCIP-1; PPP1R170; Protein kinase C inhibitor protein 1; protein phosphatase 1, regulatory subunit 170; RCJMB04_5e12; tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma; tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, gamma polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; wu:fd20c02; wu:fj53d01; ywhag; ywhag1; zgc:73131
Símbolos de los genes YWHAG
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human 14-3-3 gamma (135-186AA).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 22628, 56010, 7532
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.