missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMC4 Recombinant Rabbit Monoclonal Antibody (24GB1400)
GREENER_CHOICE

Código de producto. 30275790
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
Código de producto. Cantidad unitSize
30275790 100 μL 100 microlitros
30274803 20 μL 20 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30275790

Marca: Invitrogen™ MA557195

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

PSMC4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the ATPase subunits, a member of the triple-A family of ATPases which have a chaperone-like activity. This subunit has been shown to interact with an orphan member of the nuclear hormone receptor superfamily highly expressed in liver, and with gankyrin, a liver oncoprotein. Two transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PSMC4
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1400
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica PSMC4
N.º de referencia del gen P43686, P54775, Q63570
Alias de gen 26S protease regulatory subunit 6B; 26S proteasome AAA-ATPase subunit RPT3; 26S proteasome regulatory subunit 6B; CAR interacing protein 21; CAR interacting protein 21; CIP21; MB67-interacting protein; MGC13687; MGC23214; MGC8570; MIP224; protease 26S subunit 6; proteasome (prosome, macropain) 26S subunit, ATPase, 4; proteasome 26S ATPase subunit 4; proteasome 26S subunit ATPase 4; proteasome 26S subunit, ATPase 4; PSMC4; RPT3; S6; S6 ATPase; Tat-binding protein 7; TBP7; TBP-7
Símbolos de los genes PSMC4
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human Tbp7 (aa 350-418).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117262, 23996, 5704
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.