Learn More
Reagecon 100ML Flame Photometer Standard Sodium 210mmol/l and Potassium 140mmol/l
Marca:  Reagecon FCNK12-M
100ML Flame Photometer Standard Sodium 210mmol/l and Potassium 140mmol/l
Código de producto. 16470684
  • 95.95€ / 100 ml