missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alclofenac, Mikromol™
Click to view available options
Cantidad:
100 mg
Tamaño de la unidad:
100 miligramos
Descripción
Alclofenac
Specifications
Specifications
| Nombre del producto químico o material | Alclofenac |
| CAS | 22131-79-9 |
| Nombres de analitos o componentes | Alclofenac |
| Condiciones de envío | Room Temperature |
| Fórmula molecular | C11 H11 Cl O3 |
| Certificaciones/conformidad | ISO 17025 |
| Sinónimo | Benzeneacetic acid, 3-chloro-4-(2-propen-1-yloxy)-Acetic acid, [4-(allyloxy)-3-chlorophenyl]- (8CI), Benzeneacetic acid, 3-chloro-4-(2-propenyloxy)- (9CI), Aclofenac, Alclofenac, Alclophenac, Allopydin, Epinal, MY 101, Medifenac, Mervan, Neosten, Neoston, Prinalgin, Reufenac, W 7320, Zumaril, [3-Chloro-4-(allyloxy)phenyl]acetic acid, [4-(Allyloxy)-3-chlorophenyl]acetic acid |
| Fórmula InChI | InChI=1S/C11H11ClO3/c1-2-5-15-10-4-3-8(6-9(10)12)7-11(13)14/h2-4,6H,1,5,7H2,(H,13,14) |
| SMILES | OC(=O)Cc1ccc(OCC=C)c(Cl)c1 |
| Nombre IUPAC | 2-(3-chloro-4-prop-2-enoxyphenyl)acetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction