Learn More
Hamilton™ 100 μL 1800 Series Gas Tight Syringe
Descripción
- Attached to the flange of the syringe is a blue syringe holder, which provides an area to hold the syringe that prevents heat transfer, dispense inaccuracies, and plunger blow outs.
- Provides additional support to small-volume plungers that are prone to bending during injection.
- This series of syringes are ideal for dispensing volumes up to 100 μL.
Especificaciones
Especificaciones
| Esterilizables en autoclave | Autoclavable |
| Material | PTFE |
| Calibre de aguja | 22s G |
| Diámetro interior (imperial) | 0,057 in |
| Diámetro interior de la aguja (imperial) | 0,0059 in |
| Diámetro interior de la aguja (métrico) | 0.15 mm |
| Diámetro exterior de la aguja (imperial) | 0,0283 in |
| Diámetro exterior (imperial) | 0,305 in |
| Diámetro interior (métrico) | 1.49 mm |
| Diámetro exterior (métrico) | 7.75 mm |
| Mostrar más |